

Calcium nitrate tetrahydrate ACS
Supplier: VWR
Certificates
About this item
CAS Number:13477-34-4
:
Formula:Ca(NO₃)₂·4H₂O
MDL Number:MFCD00149604
:236.15 g/mol
Merck Index:13,01687
Melting Pt:~ 42 °C
Density:1.86 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-332-1
UN:1454
ADR:5.1,III
:GHS07
:GHS05
Specification Test Results
- Barium:< 0.005%
- Chloride:<= 0.005%
- Heavy Metals (as Pb):<= 0.0005%
- Insolubles:<= 0.005%
- Iron:<= 0.0005%
- Magnesium:<= 0.05%
- Nitrite:<= 0.001%
- pH (5%, Water) @25°C:5 - 7
- Potassium:<= 0.005%
- Purity:99% - 103%
- Sodium:<= 0.01%
- Strontium:<= 0.05%
- Sulfate:<= 0.002%