L(+)-Ornithine hydrochloride ≥99%, Reagent Grade
10118-204
: VWR
:
- Size:500 g
- Environmentally Preferable:
- Cat. No.:10118-204
- Supplier no.:0958-500G
- Ammonium:<= 0.1%
- Heavy Metals:<= 10 ppm
- Iron:<= 10 ppm
- Loss on Drying:<= 0.5%
- Purity:>= 99%
- Specific Rotation (Dried, c=4, 6N HCl):23 °- 25 °
- Sulfate:<= 0.02%
: H-Orn-OH Hydrochloride, H-Orn-OH·HCl
:
:168.62 g/mol
:221-678-6
:3184-13-2
:C₅H₁₂N₂O₂·HCl
:MFCD00064562