ContactUsLinkComponent
Flexible Brush Handle, Nylon, Remco Products
Flexible Brush Handle, Nylon, Remco Products
  76006-960
undefined
Flexible Brush Handle, Nylon, Remco Products
  76006-960
 :  53525
  • Handle material:
    Nylon
  • Cat. No.:
    76006-960
  • Handle length:
    152.4 cm (60)"
  • Handle color:
    White
This nylon flexible handle is used with a tube brush for cleaning around sensitive areas such as glass, and its flexibility allows it to follow curved pipes.
Weight: 0.2 lbs.