F.C. Bond Ball Mill, Bico
20058-110
: Bico
- Description:3 Phase, 60 Cycle
- Environmentally Preferable:
- Cat. No.:20058-110
- Electrical:220/440 V
- Drum size:12×12"
- Supplier no.:395-50
Designed by F.C. Bond for use by cement companies and quality control laboratories throughout the world. Can be used in calculating the grindability of all ore.
: 285 Ball Charge. Contact your Avantor sales representative for more information.