Order Entry
Puerto Rico
ContactUsLinkComponent
P3 SureLight® molecular biology stain
P3 SureLight® molecular biology stain
Catalog # 10065-688
CAS Number:  
undefined
P3 SureLight® molecular biology stain
Catalog # 10065-688
Supplier Number:  D7-000
CAS Number:  

Specifications

  • Size:
    5 mg
  • Cat. No.:
    10065-688
  • Supplier no.:
    D7-000

Specifications

About this item

 :
Storage Temperature:Refrigerator