Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Myd88 Fragment (from E. coli)
Human Recombinant Myd88 Fragment (from E. coli)
Catalog # 10073-126
Supplier:  Prosci
CAS Number:  
undefined
Human Recombinant Myd88 Fragment (from E. coli)
Catalog # 10073-126
Supplier:  Prosci
Supplier Number:  95-104
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Protein Synonyms:
    MyD88 MYD88D
  • Protein/Peptide Name:
    Myd88 Fragment
  • Purity:
    >95%
  • Molecular Weight:
    15 kDa (Calculated)
  • Formulation:
    Liquid
  • Tested Applications:
    ELISA, Western Blot
  • Cat. No.:
    10073-126
  • Supplier no.:
    95-104

Specifications

About this item

The pro-inflammatory cytokine IL-1 induced cellular response requires IL-1 receptor complex including IL-1RI and IL-1RAcP. MyD88 has been identified as an adapter molecule in the IL-1 signaling pathway (1). MyD88 associates with and recruits IRAK to the IL-1 receptor complex in response to IL-1 treatment and dominant negative form of MyD88 attenuates IL-1R-mediated NF-B activation. MyD88 is also employed as a regulator molecule by IL-18 receptor and human Toll receptor (2,3), which are members in the Toll/IL-1R family of receptors. Targeted disruption of the MyD88 gene results in lose of cellular responses to IL-1 and IL-18, and MyD88-deficient mice lack responses to bacterial product LPS that employs Toll-like receptors 2 and 4 (TLR2 and TLR4) as the signaling receptors. MyD88 is a general adapter protein for the Toll/IL-1R family of receptors and plays an important role in the inflammatory response induced by cytokines IL-1 and IL-18 and endotoxin. MyD88 gene is expressed in many tissues.

This recombinant protein can be used for WB, ELISA, MS and neutralization assays. For research use only.

100mM sodium phosphate, 500mM NaCl, 25 mM imidazole, 10% gycerol.

Store in working aliquots at -80°C. Avoid freeze/thaw cycles.