Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Osteonectin (from CHO Cells)
Human Recombinant Osteonectin (from CHO Cells)
Catalog # 10073-048
Supplier:  Prosci
CAS Number:  
Human Recombinant Osteonectin (from CHO Cells)
Catalog # 10073-048
Supplier:  Prosci
Supplier Number:  40-571
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Size:
    0.01 mg
  • Biological Activity:
    Biological activity was determined by its ability to increase alkaline phosphatase activity in differentiating MC3T3 cells using a concentration of 0.5-0.7 μg/mL. Endotoxin level is less than 0.1 ng per μg (1EU/μg).
  • Protein Synonyms:
    Basement-membrane protein 40|ON|SPARC|BM-40
  • Protein/Peptide Name:
    Osteonectin
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. No.:
    10073-048
  • Supplier no.:
    40-571

Specifications

About this item

SPARC/Osteonectin is a secreted, evolutionarily conserved collagen-binding glycoprotein that is involved in a variety of cellular activities. It is highly expressed in tissues undergoing morphogenesis, remodeling and wound repair. SPARC/Osteonectin and its related peptides bind to numerous proteins of the extracellular matrix (ECM), affect ECM protein expression, influence cellular adhesion and migration, and modulate growth factor-induced cell proliferation and angiogenesis. SPARC/Osteonectin consists of three domains; an N-terminal acidic region that binds calcium ions with low affinity, a module containing two EF-hand motifs that bind calcium with high affinity, and a cysteine-rich follistatin-like domain. Recombinant human SPARC/Osteonectin is a glycoprotein containing 286 amino acids that migrates at an apparent MW of 43.7 kDa by SDS-PAGE analysis due to the effect of glycosylation.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.