Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Enterokinase (from CHO cells)
Human Recombinant Enterokinase (from CHO cells)
Catalog # 10073-020
Supplier:  Prosci
CAS Number:  
Human Recombinant Enterokinase (from CHO cells)
Catalog # 10073-020
Supplier:  Prosci
Supplier Number:  40-552
CAS Number:  

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Size:
    0.01 mg
  • Form:
    Lyophilized
  • Enzyme Name:
    Enterokinase
  • Enzyme Activity:
    Sequentially cleaves carboxyl side of D-D-D-D-K. Endotoxin level is less than 0.2 ng per μg (2EU/μg).
  • Enzyme Synonyms:
    ENTK, PRSS7, ENTK, Enteropeptidase, Enterokinase
  • Purity:
    >90%
  • Cat. No.:
    10073-020
  • Supplier no.:
    40-552

Specifications

About this item

Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Most proteases act in a specific manner, hydrolyzing bonds at or adjacent to specific residues or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes including prenatal and postnatal development, reproduction, signal transduction, the immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, frequently used in the analysis and production of proteins. Enterokinase sequentially cleaves carboxyl side of D-D-D-D-K. Human Enterokinase is expressed as a linear 1019 amino acid polypeptide precursor glycoprotein. Proteolytic processing of this precursor generates the biologically active form of Enterokinase, which consists of two polypeptide chains (heavy chain and light chain) held together by a single disulfide bond, resulting in formation of a biologically active heterodimer. The heavy chain consists of 784 amino acid residues, and the light consists of 235 amino acid residues.