About this item
The attractive design and sturdy construction of the Secador® line of desiccator cabinets assures easy and reliable dry storage of moisture- and UV-sensitive products.
- Sturdy, reliable dry storage
- Protects moisture- and UV-sensitive products
- Removable, perforated shelves facilitate dry air circulation
- Ergonomic carrying handle for easy transportation
These units include a built-in suitcase style carrying handle for easy transportation
Polycarbonate construction features a UV light blocker and is resistant to staining, crazing, and chemical attack. The large door maximizes access to interior space and has a patented seal design, tab latches and security loops for a padlock or tamper-evident seals. An easy-to-view dial hygrometer is located on the pull down ‘oven style’ door.
Two removable, perforated shelves facilitate circulation of dry air and allow space for desiccant under the bottom shelf. U.S. Patents: 6,834,920; 7,318,630 and D474,626.
Specifications
- Catalog No:
- 47743-426
- Ext. Dimensions:
- 21.3W×41.4D×37.3H cm (8.4×16.3×13.4”)
- Int. Volume:
- 19.8 L (0.7 cu.ft.)