Order Entry
Puerto Rico
ContactUsLinkComponent
MS Reciprocal Shaker, MAJOR SCIENCE BE
MS Reciprocal Shaker, MAJOR SCIENCE BE
Catalog # 490016-426
Supplier:  MAJOR SCIENCE BE
MS Reciprocal Shaker, MAJOR SCIENCE BE
Catalog # 490016-426
Supplier:  MAJOR SCIENCE BE
Supplier Number:  MS-NRC-30

Specifications

  • Description:
    Shaker with Flat Non-Slip Rubber Mat
  • Cat. No.:
    490016-426
  • Electrical:
    110/220 V
  • Platform Size:
    30×30 cm
  • Supplier no.:
    MS-NRC-30

Specifications

About this item

The Shaker features a left to right reciprocal shaking, providing gentle to vigorous shaking for a variety of applications. The operation may be continuous or timed, with the integral electronic timer ensuring accurate repeatability of time-sensitive incubations.

  • Continuous or timed operation with automatic stop
  • Variable shaking speed form 5 to 100 rpm
  • Interchangeable/stacking platforms, and accessories for a variety of vessels
  • 15 kg capability able to support a small water bath