Order Entry
Puerto Rico
ContactUsLinkComponent
 
Small Intestine Lysate, ProSci
 :  Prosci
 
Small Intestine Lysate, ProSci
 :  Prosci

 

  • Storage Conditions:
    Store at 2-8°C for continuous use. For extended storage, freeze working aliquots at -70°C.
  • Storage Buffer:
    Small Intestine lysate is supplied in SDS sample buffer containing 5% β-mercaptoethanol.
  • Concentration:
    2 mg/mL
  • Tested Applications:
    WB

 

Frequently Bought Together