Order Entry
Puerto Rico
ContactUsLinkComponent
Liver Lysate (14 Days Old), ProSci Inc
Liver Lysate (14 Days Old), ProSci Inc
Catalog # 10062-392
Supplier:  Prosci
CAS Number:  
Liver Lysate (14 Days Old), ProSci Inc
Catalog # 10062-392
Supplier:  Prosci
Supplier Number:  1404-14
CAS Number:  

Specifications

  • Cell Format:
    Cryopreserved
  • Description:
    Normal
  • Size:
    0.1 mg
  • Storage Conditions:
    Store at 2-8°C for continuous use. For extended storage, freeze working aliquots at -70°C.
  • Storage Buffer:
    Liver lysate (14 Days Old) is supplied in SDS sample buffer containing 5% β-mercaptoethanol.
  • Concentration:
    2 mg/mL
  • Tested Applications:
    WB
  • Cat. No.:
    10062-392
  • Source:
    Mouse
  • Tissue:
    Liver
  • Supplier no.:
    1404-14

Specifications

About this item

Liver tissue lysate (14 Days Old) was prepared by homogenization in modified RIPA buffer (150 mM sodium chloride, 50 mM Tris-HCl, pH 7.4, 1 mM ethylenediaminetetraacetic acid, 1 mM phenylmethylsulfonyl fluoride, 1% Triton X-100, 1% sodium deoxycholic acid, 0.1% sodium dodecylsulfate, 5 μg/ml of aprotinin, 5 μg/ml of leupeptin

Tissue and cell debris was removed by centrifugation. Protein concentration was determined with Bio-Rad protein assay. The product was boiled for 5 min in 1 x SDS sample buffer (50 mM Tris-HCl pH 6.8, 12.5% glycerol, 1% sodium dodecylsulfate, 0.01% bromophenol blue) containing 5% β-mercaptoethanol.