VWR® Life Science’s Uracil-DNA Glycosylase (UNG), Cod is a thermolabile recombinant enzyme produced in E
- Complete, irreversible heat-inactivation
- Prevents carryover contamination in PCR, qPCR, RT-PCR, and qRT-PCR
- Works with all commercially available master mixes
- Only a 5 minute incubation step is required prior to amplification
In contrast to competing Uracil-DNA Glycosylases, VWR® arctic-derived UNG has the distinct advantage of having the low inactivation temperature of 45°C, which enables carryover decontamination of one-step reverse transcription reactions with UNG prior to the cDNA synthesis step. Inactivation is irreversible after heating just 20 minutes at 55°C or 1 second at 95°C, allowing for stability of the cDNA that has incorporated dUTP. The irreversible inactivation feature of VWR® UNG also facilitates greater downstream manipulation and stability of newly synthesized dUTP-containing DNA by PCR, allowing for long-term storage, restriction digestion, cloning, sequencing and hybridization applications. All amplification reactions must use dUTP-containing dNTP mixtures in order for the UNG decontamination method to be effective.
Storage/Stability
Product is stable at least 2 years when stored frozen (-20 – 0°C). Product may be stored cold (4 – 8°C) for up to 6 months.
coli (ung-) by a modified ung gene derived from Atlantic Cod. It degrades uracil-containing single-stranded and double-stranded DNA, but not RNA or thymidine-containing DNA, by hydrolyzing the N-glycosidic bond between deoxyribose sugar and the base in uracil. This generates alkaline sensitive apyramidinic sites in the DNA that will be cleaved upon a combination of alkaline conditions and high temperature. Pretreatment of samples with UNG prevents PCR carryover contamination in labs that substitute dUTP in place of dTTP during all amplification reactions. PCR products containing uracil become substrates for UNG and will be degraded if they are present in subsequent reaction mixtures subjected to UNG treatment. Only DNA templates containing thymidine are not degraded by the treatment and will be amplified.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Enzyme Name:Uracil-DNA Glycosylase