ContactUsLinkComponent
Pleated (Fluted) Filter Paper, Grade 542, Ahlstrom
Pleated (Fluted) Filter Paper, Grade 542, Ahlstrom
Supplier:  Ahlstrom
Certificates
undefined
Pleated (Fluted) Filter Paper, Grade 542, Ahlstrom
Supplier:  Ahlstrom
Specifications
  • Catalog No:
  • 38002-066
  • 38002-044
  • 47749-636
  • 47749-638
  • 47749-634
  • 38002-088
  • Flow Rate:
  • Slow
  • Slow
  • Slow
  • Slow
  • Slow
  • Slow
  • Load Capacity:
  • Medium
  • Medium
  • Medium
  • Medium
  • Medium
  • Medium
  • Retention:
  • 2.0 µm
  • 2.0 µm
  • 2.0 µm
  • 2.0 µm
  • 2.0 µm
  • 2.0 µm