Order Entry
Puerto Rico
ContactUsLinkComponent
Rat Recombinant IL5 (from E. coli)
Rat Recombinant IL5 (from E. coli)
Supplier:  Prosci
Certificates
Rat Recombinant IL5 (from E. coli)
Supplier:  Prosci

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Biological Activity:
    The ED50 was determined by the dose - dependent stimulation of the proliferation of human TF - 1 cells is < 2.0 ng/mL, corresponding to a specific activity of > 5 x 105 units/mg.
  • Protein Synonyms:
    Interleukin-5|EDF|TRF|IL-5|B-cell differentiation factor I
  • Protein/Peptide Name:
    IL5
  • Purity:
    >98%
  • Formulation:
    Lyophilized

Specifications

Related Information