Human Recombinant BLC (from E. coli)
Catalog # 10072-176
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.005 mg
- Biological Activity:Biological activity was determined by its ability to chemoattract human B cells using a concentration range of 1.0 - 10.0 ng/mL.
- Protein Synonyms:Angie|BLC|ANGIE|BLR1L|BCA1|BCA-1|ANGIE2|SCYB13|C-X-C motif chemokine 13
- Protein/Peptide Name:BLC
- Purity:>98%
- Formulation:Lyophilized
- Cat. No.:10072-176
- Supplier no.:40-399
Specifications
About this item
BCA-1/BLC, a CXC chemokine, is expressed in the liver, spleen, lymph nodes, appendix and stomach. It exerts its activities through its only receptor CXCR5. BCA-1/BLC is a potent chemoattractant for B lymphocytes and induces weak chemotactic response in T cells and macrophages. It manifests no activity on neutrophils and monocytes. Recombinant Human BCA-1 is a 10.3 kDa protein containing 87 amino acid residues including the four highly conserved cysteine residues present in CXC chemokines.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.