Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant APRIL (from E. coli)
Mouse Recombinant APRIL (from E. coli)
Catalog # 10072-462
Supplier:  Prosci
CAS Number:  
Mouse Recombinant APRIL (from E. coli)
Catalog # 10072-462
Supplier:  Prosci
Supplier Number:  40-453
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    0.005 mg
  • Biological Activity:
    Biological activity was measured by its ability to induce cell proliferation of activated T cells.
  • Protein Synonyms:
    Tumor necrosis factor ligand superfamily member 13|A proliferation-inducing ligand|APRIL|April|Trdl1|2310026N09Rik|Tall2
  • Protein/Peptide Name:
    APRIL
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. No.:
    10072-462
  • Supplier no.:
    40-453

Specifications

About this item

APRIL, a member of the TNF superfamily, is expressed in monocytes, macrophages, certain transformed cell lines, certain cancers of colon, and lymphoid tissues. APRIL, along with another TNF family member, BAFF, compete for two receptors, TACI and BCMA. APRIL has the ability to stimulate proliferation of various tumor cell lines including Jurkat T cells and MCF-7 carcinoma cells. Like BAFF, APRIL also stimulates the proliferation of B and T cells. The human APRIL gene codes for at least four alternatively spliced transcriptional variants, which give rise to different isoforms of the APRIL precursor protein. All isoforms can be cleaved by the protease, furin, to release a soluble C-terminal fragment, which comprises the TNF like receptor binding of the APRIL precursor. Recombinant murine APRIL is a soluble 21.9 kDa protein, consisting of 192 amino acid residues.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.