Human Recombinant MIP5 (from E. coli)
Catalog # 10072-996
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.005 mg
- Biological Activity:Biological activity was determined by its ability to chemoattract human T lymphocytes using a concentration range of 1.0 - 10.0 ng/mL.
- Gene ID:6359
- Protein Synonyms:MIP-1 delta|MIP-5|LKN-1|HMRP-2B|SCYA15|MRP-2B|Chemokine CC-2|MIP5|LKN1|HCC-2|MIP-1D|C-C motif chemokine 15|SY15|NCC3|NCC-3|SCYL3
- Protein/Peptide Name:MIP5
- Purity:>98%
- Formulation:Lyophilized
- Cat. No.:10072-996
- Supplier no.:40-395
Specifications
About this item
MIP-5 is a CC chemokine that is expressed in the heart, skeletal muscle and adrenal gland. MIP-5 primarily signals through he CCR1 receptor, but also has been found to bind to CCR3. MIP-5 is chemotactic towards T cells and monocytes. Recombinant human MIP-5 is a 10.1 kDa protein containing 92 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.