Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant CXCL16 (from E. coli)
Mouse Recombinant CXCL16 (from E. coli)
Catalog # 10072-964
Supplier:  Prosci
CAS Number:  
Mouse Recombinant CXCL16 (from E. coli)
Catalog # 10072-964
Supplier:  Prosci
Supplier Number:  40-355
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    0.005 mg
  • Biological Activity:
    Biological activity was determined by its ability to chemoattract murine lymphocytes using a concentration of 20 - 1000 ng/mL.
  • Protein Synonyms:
    SR-PSOX|BB024863|0910001K24Rik|b2b498Clo|Zmynd15|AV290116|CXCL16v1|C-X-C motif chemokine 16|CXCL16v2|Srpsox|Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein
  • Protein/Peptide Name:
    CXCL16
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. No.:
    10072-964
  • Supplier no.:
    40-355

Specifications

About this item

CXCL16 is a member of the CXC chemokine family and signals through the CXCR6 receptor. CXCL16 may play a role in attracting lymphocyte subsets during inflammation and may facilitate certain immune responses. The chemokine domain of CXCL16 contains six cysteine residues including the four highly conserved cysteine residues characteristic of CXC chemokines. The CXCL16 gene codes for a 273 amino acid polypeptide, which includes a 29 amino acid cytoplasmic domain and transmembrane sequence containing approximately 20 amino acids. The extracellular portion of CXCL16 contains a chemokines domain and an extended C-terminal ?mucin-like stalk? sequence. The extracellular domain contains 89 amino acid residues (86 a.a. residues for the murine homolog). Recombinant murine CXCL16 is a 9.9 kDa protein containing 88 amino acid residues.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.