Mouse Recombinant SDF1 alpha (from E. coli)
Catalog # 10072-114
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Size:0.002 mg
- Biological Activity:Biological activity was determined by its ability to chemoattract human peripheral blood monocytes using a concentration of 50 - 100 ng/mL.
- Protein Synonyms:Scyb12|12-O-tetradecanoylphorbol 13-acetate repressed protein 1|Tpar1|Stromal cell-derived factor 1|Pbsf|SDF-1|Tlsf|Sdf1
- Protein/Peptide Name:SDF1 alpha
- Purity:>98%
- Formulation:Lyophilized
- Cat. No.:10072-114
- Supplier no.:40-347
Specifications
About this item
SDF-1 alpha and beta are stromal derived CXC chemokines, and signal through the CXCR4 receptor. SDF-1alpha and beta chemoattract B and T cells, and have been shown to induce migration of CD34+ stem cells. Additionally, the SDF-1 proteins exert HIV suppressive activity in cells expressing the CXCR4 receptor. Recombinant murine SDF-1alpha is a 7.9 kDa protein containing 68 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.