Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant LIX (from E. coli)
Mouse Recombinant LIX (from E. coli)
Catalog # 10072-110
Supplier:  Prosci
CAS Number:  
Mouse Recombinant LIX (from E. coli)
Catalog # 10072-110
Supplier:  Prosci
Supplier Number:  40-345
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    0.005 mg
  • Biological Activity:
    Biological activity was determined by its ability to chemoattract human neutrophils using a concentration range of 10 - 100 ng/mL.
  • Protein Synonyms:
    Scyb5|Cytokine LIX|ENA-78|AMCF-II|Scyb6|LIX|GCP-2|C-X-C motif chemokine 5
  • Protein/Peptide Name:
    LIX
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Cat. No.:
    10072-110
  • Supplier no.:
    40-345

Specifications

About this item

The murine homolog of ENA-78 is called LIX. ENA-78/LIX is a CXC chemokine that signals through the CXCR2 receptor. It is expressed in monocytes, platelets, endothelial cells, and mast cells. ENA-78/LIX is a chemoattractant for neutrophils. The three naturally occurring variants of human ENA-78; ENA 5-78, ENA 9-78 and ENA 10-78, contain 74, 70, and 69 amino acid residues, respectively, and possess the same biological activity. ENA-78/LIX contains the four conserved cysteine residues present in CXC chemokines, and also contains the 'ELR' motif common to CXC chemokine that bind to the CXCR1 and CXCR2 receptors. Recombinant murine LIX is a 9.8 kDa protein consisting of 93 amino acid residues.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.