Human Recombinant Tdo2 (from E. coli)
Catalog # 10072-356
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Protein Synonyms:TRPO|TO|TDO|TPH2|Tryptophan 2|Tryptamin 2|3-dioxygenase
- Protein/Peptide Name:Tdo2
- Purity:>95%
- Molecular Weight:46.7 kDa (Calculated)
- Tested Applications:ELISA, Western Blot, MS
- Cat. No.:10072-356
- Supplier no.:XW-RP3263
Specifications
About this item
The tetramer binds two molecules of protoheme IV. It is a homotetramer involved in tryptophan catabolism. There is a broad specificity towards tryptamine and derivatives including D- and L-tryptophan, 5-hydroxytryptophan and serotonin.
- Store in working aliquots at –80 °C
- Avoid freeze/thaw cycles
This recombinant protein can be used for WB, ELISA, MS.
10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN