Human Recombinant Snx5 (from E. coli)
Catalog # 10072-350
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Protein Synonyms:Sorting nexin-5
- Protein/Peptide Name:Snx5
- Purity:>95%
- Molecular Weight:46.5 kDa (Calculated)
- Tested Applications:ELISA, Western Blot, MS
- Cat. No.:10072-350
- Supplier no.:XW-RP3253
Specifications
About this item
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein binds to fanconi anemia complementation group A protein, but its function is unknown. This gene results in two transcript variants encoding the same protein.
This recombinant protein can be used for WB, ELISA, MS.
10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN, 10mM DTT
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.