Human Recombinant Pxf (from E. coli)
Catalog # 10072-340
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Protein Synonyms:D1S2223E|PBD12A|PXMP1|OK/SW-cl.22|33 kDa housekeeping protein|PMPI|HK33|PXF|PMP1|Peroxisomal biogenesis factor 19
- Protein/Peptide Name:Pxf
- Purity:>95%
- Molecular Weight:34.4 kDa (Calculated)
- Tested Applications:ELISA, Western Blot, MS
- Cat. No.:10072-340
- Supplier no.:XW-RP3215
Specifications
About this item
The covalent attachment of prenyl lipids, such as farnesyl or geranylgeranyl groups, by specific transferases is indispensable for the cellular sorting of many proteins. James et al. (1994) identified in hamster a farnesylated protein, called peroxisomal farnesylated protein or PxF, that localized to the outer surface of peroxisomes.
This recombinant protein can be used for WB, ELISA, MS.
10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN, 10mM DTT
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.