Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Psmc6 (from E. coli)
Human Recombinant Psmc6 (from E. coli)
Catalog # 10072-338
Supplier:  Prosci
CAS Number:  
undefined
Human Recombinant Psmc6 (from E. coli)
Catalog # 10072-338
Supplier:  Prosci
Supplier Number:  XW-RP3212
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Protein Synonyms:
    26S protease regulatory subunit 10B|SUG2|P44|26S proteasome AAA-ATPase subunit RPT4|p42|CADP44|HEL-S-73
  • Protein/Peptide Name:
    Psmc6
  • Purity:
    >95%
  • Molecular Weight:
    44.7 kDa (Calculated)
  • Tested Applications:
    ELISA, Western Blot, MS
  • Cat. No.:
    10072-338
  • Supplier no.:
    XW-RP3212

Specifications

About this item

The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 α subunits and 2 rings are composed of 7 β subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides.

This recombinant protein can be used for WB, ELISA, MS.

10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN, 10mM DTT

Store in working aliquots at -80°C. Avoid freeze/thaw cycles.