Human Recombinant PPIL1 (from E. coli)
Catalog # 10072-330
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Protein Synonyms:UNQ2425/PRO4984|hCyPX|Peptidyl-prolyl cis-trans isomerase-like 1|CGI-124|CYPL1|PPIase|Rotamase PPIL1
- Protein/Peptide Name:PPIL1
- Purity:>95%
- Molecular Weight:19.1 kDa (Calculated)
- Tested Applications:ELISA, Western Blot, MS
- Cat. No.:10072-330
- Supplier no.:XW-RP3204
Specifications
About this item
This protein is a member of the cyclophilin family of peptidylprolyl isomerases (PPIases). The cyclophilins are a highly conserved, ubiquitous family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions. Based on similarity to other PPIases, this protein could accelerate the folding of proteins and might catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.
This recombinant protein can be used for WB, ELISA, MS.
10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.