Human Recombinant Psat1 (from E. coli)
Catalog # 10072-334
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.1 mg
- Protein Synonyms:PSA|EPIP|Phosphohydroxythreonine aminotransferase|PSAT|Phosphoserine aminotransferase
- Protein/Peptide Name:Psat1
- Purity:>95%
- Molecular Weight:42.6 kDa (Calculated)
- Tested Applications:ELISA, Western Blot, MS
- Cat. No.:10072-334
- Supplier no.:XW-RP3208
Specifications
About this item
PSAT1 is likely a phosphoserine aminotransferase, based on similarity to proteins in mouse, rabbit, and Drosophila. It belongs to class-V of pyridoxal-phosphate-dependent aminotransferases and is required both in major phosphorylated pathway of serine biosynthesis and in the biosynthesis of pyridoxine.
This recombinant protein can be used for WB, ELISA, MS.
10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.