Human Recombinant Mat2A (from E. coli)
Catalog # 10072-312
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Protein Synonyms:SAMS2|Methionine adenosyltransferase 2|AMS2|S-adenosylmethionine synthase isoform type-2|MATA2|MATII|AdoMet synthase 2
- Protein/Peptide Name:Mat2A
- Purity:>95%
- Molecular Weight:46 kDa (Calculated)
- Tested Applications:ELISA, Western Blot, MS
- Cat. No.:10072-312
- Supplier no.:XW-RP3143
Specifications
About this item
Methionine adenosyltransferase (EC 2.5.1.6) catalyzes the biosynthesis of S-adenosylmethionine (AdoMet) from methionine and ATP. AdoMet is the major methyl donor for many of the transmethylation reactions in the body. The MAT2A promoter is highly GC rich, especially in the first 300 bp. The promoter contains several potential SP1-, v-myb-, and GATA-binding sites, as well as potential binding sites for C/EBP, HSF2, c-myb, and STATx. It can efficiently drive expression from a reporter gene in both Jurkat and 293 cells.
This recombinant protein can be used for WB, ELISA, MS.
10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.