Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant LXN (from E. coli)
Human Recombinant LXN (from E. coli)
Catalog # 10072-310
Supplier:  Prosci
CAS Number:  
undefined
Human Recombinant LXN (from E. coli)
Catalog # 10072-310
Supplier:  Prosci
Supplier Number:  XW-RP3139
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Protein Synonyms:
    Endogenous carboxypeptidase inhibitor|ECI|TCI|Latexin
  • Protein/Peptide Name:
    LXN
  • Purity:
    >95%
  • Molecular Weight:
    25.5 kDa (Calculated)
  • Tested Applications:
    ELISA, Western Blot, MS
  • Cat. No.:
    10072-310
  • Supplier no.:
    XW-RP3139

Specifications

About this item

Strongly similar to rat latexin (TCI, Rn.11404); may be a tissue carboxypeptidase inhibitor.

This recombinant protein can be used for WB, ELISA, MS.

10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN, 10mM DTT

Store in working aliquots at -80°C. Avoid freeze/thaw cycles.