Human Recombinant Esd (from E. coli)
Catalog # 10072-294
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Protein Synonyms:S-formylglutathione hydrolase|Esterase D|FGH
- Protein/Peptide Name:Esd
- Purity:>95%
- Molecular Weight:32.4 kDa (Calculated)
- Tested Applications:ELISA, Western Blot, MS
- Cat. No.:10072-294
- Supplier no.:XW-RP3079
Specifications
About this item
A carboxylic ester + H2O = an alcohol + a carboxylic anion. Homodimer. Cytoplasmic vesicles. There are two major electrophoretic isotypes. The sequence of the ESD*1 variant is shown. Similar to yeast HRE299.
This recombinant protein can be used for WB, ELISA, MS.
10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN, 10mM DTT
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.