Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Dnal4 (from E. coli)
Human Recombinant Dnal4 (from E. coli)
Catalog # 10072-288
Supplier:  Prosci
CAS Number:  
undefined
Human Recombinant Dnal4 (from E. coli)
Catalog # 10072-288
Supplier:  Prosci
Supplier Number:  XW-RP3065
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.1 mg
  • Protein Synonyms:
    Dynein light chain 4|PIG27|axonemal
  • Protein/Peptide Name:
    Dnal4
  • Purity:
    >90%
  • Molecular Weight:
    12.1 kDa (Calculated)
  • Tested Applications:
    ELISA, Western Blot, MS
  • Cat. No.:
    10072-288
  • Supplier no.:
    XW-RP3065

Specifications

About this item

Force generating protein of respiratory cilia. Produces force towards the minus ends of microtubules.

This recombinant protein can be used for WB, ELISA, MS.

10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN, 10mM DTT

Store in working aliquots at -80°C. Avoid freeze/thaw cycles.