Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Atp6V1E1 (from E. coli)
Human Recombinant Atp6V1E1 (from E. coli)
Catalog # 10072-270
Supplier:  Prosci
CAS Number:  
undefined
Human Recombinant Atp6V1E1 (from E. coli)
Catalog # 10072-270
Supplier:  Prosci
Supplier Number:  XW-RP3020
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Protein Synonyms:
    ATP6V1E|Vma4|ATP6E2|ATP6E|P31|V-type proton ATPase subunit E 1|V-ATPase 31 kDa subunit|V-ATPase subunit E 1
  • Protein/Peptide Name:
    Atp6V1E1
  • Purity:
    >95%
  • Molecular Weight:
    26 kDa (Calculated)
  • Tested Applications:
    ELISA, Western Blot, MS
  • Cat. No.:
    10072-270
  • Supplier no.:
    XW-RP3020

Specifications

About this item

This protein is a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, and receptor-mediated endocytosis. V-ATPase is comprised of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of a hexamer of three A and three B subunits plus the C, D, and E subunits. It contains the ATP catalytic site. The protein is known as the E subunit and is found ubiquitously.

This recombinant protein can be used for WB, ELISA, MS.

10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN, 10mM DTT

Store in working aliquots at -80°C. Avoid freeze/thaw cycles.