Human Recombinant Khk (from E. coli)
Catalog # 10072-306
Supplier: Prosci
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Protein Synonyms:Hepatic fructokinase|Ketohexokinase
- Protein/Peptide Name:Khk
- Purity:>90%
- Molecular Weight:34.3 kDa (Calculated)
- Tested Applications:ELISA, Western Blot, MS
- Cat. No.:10072-306
- Supplier no.:XW-RP3126
Specifications
About this item
KHK encodes the gene ketohexokinase that catalyzes conversion of fructose to fructose-1-phosphate. The splice variant presented encodes the highly active form found in liver, renal cortex, and small intestine, while the alternate variant encodes the lower activity form found in most other tissues.
This recombinant protein can be used for WB, ELISA, MS.
10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN, 10mM DTT
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.