Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant Dr6 Cytoplasmic Domain (from E. coli)
Mouse Recombinant Dr6 Cytoplasmic Domain (from E. coli)
Catalog # 10073-150
Supplier:  Prosci
CAS Number:  
undefined
Mouse Recombinant Dr6 Cytoplasmic Domain (from E. coli)
Catalog # 10073-150
Supplier:  Prosci
Supplier Number:  95-120
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Protein Synonyms:
    Tumor necrosis factor receptor superfamily member 21|DR6 DR6|Death receptor 6|UNQ437/PRO868|DR6|CD358|BM-018
  • Protein/Peptide Name:
    Dr6 Cytoplasmic Domain
  • Purity:
    >95%
  • Molecular Weight:
    36 kDa (Calculated)
  • Formulation:
    Liquid
  • Tested Applications:
    ELISA, Western Blot
  • Cat. No.:
    10073-150
  • Supplier no.:
    95-120

Specifications

About this item

Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Disruption of this process has been implicated in a variety of diseases such as cancer (reviewed in 1). Apoptosis is induced by certain cytokines including TNF and Fas ligand of the TNF family through their death domain containing receptors, TNF-R1 and Fas. Several death receptors including DR3, DR4, DR5, and DR6 have been identified. Like TNF-R1, DR6 (also known as TNFRSF21) interacts with death domain containing adapter molecule TRADD. Overexpression of DR6 induces apoptosis and activates NF-B and JNK. DR6 is widely expressed in human tissues and cell lines (2). Recently, DR6 was found to interact with an amino-terminal fragment of the Beta-amyloid protein (APP) in neurons, activating a caspase 6-dependent apoptotic event leading to axonal degeneration and pruning (3).

This recombinant protein can be used for WB, ELISA, MS and neutralization assays. For research use only.

1X PBS containing 10% glycerol

Store in working aliquots at -80°C. Avoid freeze/thaw cycles.