Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant MTCH2 (from E. coli)
Mouse Recombinant MTCH2 (from E. coli)
Catalog # 10073-146
Supplier:  Prosci
CAS Number:  
undefined
Mouse Recombinant MTCH2 (from E. coli)
Catalog # 10073-146
Supplier:  Prosci
Supplier Number:  95-118
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Protein Synonyms:
    2310034D24Rik|HSPC032|Mitochondrial carrier homolog 2|4930539J07Rik
  • Protein/Peptide Name:
    MTCH2
  • Purity:
    >95%
  • Molecular Weight:
    19 kDa (Calculated)
  • Formulation:
    Liquid
  • Tested Applications:
    ELISA, Western Blot
  • Cat. No.:
    10073-146
  • Supplier no.:
    95-118

Specifications

About this item

Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Disruption of this process has been implicated in a variety of diseases such as cancer (reviewed in 1). The Bcl-2 family of proteins is comprised of critical regulators of apoptosis that can be divided into two classes: those that inhibit apoptosis and those that promote cell death (reviewed in 2 and 3). MTCH2 is a member of the mitochondrial carrier protein family that catalyze the exchange of substrates across the inner mitochondrial membrane and is targeted by Bid, a pro-apoptotic Bcl-2 family member, in response to apoptotic signals (4-6), suggesting that MTCH2 may play a key role in the mitochondrial apoptosis pathway. The doublet observed in the SDS-PAGE is likely due to incomplete removal of the signal sequence.

This recombinant protein can be used for WB, ELISA, MS and neutralization assays. For research use only.

1X PBS containing 10% glycerol

Store in working aliquots at -80°C. Avoid freeze/thaw cycles.