Vikan® Dish Brushes with Scraper, Remco Products
Supplier: Remco Products
Certificates
About this item
The head of this fully color-coded dish brush has angled bristles which reaches perfectly into corners
- Block material is polypropylene; bristle material is polyester
- With small scraper at the tip to loosen stubborn dirt
- Dishwasher safe
- Autoclavable up to 250°F
The raised angle of the grip promotes quick and effective cleaning because of the operator’s hand and arm are in the ergonomically correct position.
Width: 2.4''
Height: 2''
Certifications: FDA compliant material.