Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human Recombinant TGF alpha (from <i>E. coli</i>)

Human Recombinant TGF alpha (from E. coli)

0.02 mg
Catalog # 10072-624
Supplier:   Prosci
Supplier #:  40-117
Human Recombinant TGF alpha (from <i>E. coli</i>)
Catalog # 10072-624
Supplier:  Prosci
CAS Number:  
Human Recombinant TGF alpha (from E. coli)

0.02 mg

CAS Number:  
Price
$323.00
Log in for availability
Each (20µG)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

TGF-α is an EGF-related polypeptide growth factor that signals through the EGF receptor, and stimulates the proliferation of a wide range of epidermal and epithelial cells. It is produced by monocytes, keratinocytes, and various tumor cells. TGF-α induces transformation anchorage independence in cultured cells. Human, murine and rat TGF-α are cross-species reactive. Recombinant human TGF-α is a 50 amino acid polypeptide (5.5 kDa) which shares approximately 40% sequence homology with EGF, including 6 conserved cysteine residues, which form 3 intramolecular disulfide bonds.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Documents

Specifications

Specifications

  • Size:
    0.02 mg
  • Cat. No.:
    10072-624
  • Biological Activity:
    The ED50 was determined by the dose - dependent stimulation of thymidine uptake by BALB/c 3T3 cells is ≤ 0.2 ng/mL, corresponding to a specific activity of ≥ 5 x 10<sup>6</sup> units/mg.
  • Protein/Peptide Type:
    Recombinant
  • Formulation:
    Lyophilized
  • Purity:
    >98%
  • Protein Synonyms:
    Tgf-alpha
  • Protein/Peptide Name:
    TGF alpha
  • Species:
    Human
  • Source:
    E. coli

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.