Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant TGF beta 1 (from HEK293 Cells)
Human Recombinant TGF beta 1 (from HEK293 Cells)
Supplier:  Prosci
Certificates
Human Recombinant TGF beta 1 (from HEK293 Cells)
Supplier:  Prosci

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Biological Activity:
    The ED50 was determined by TGF - beta1's ability to inhibit the mouse IL - 4 - dependent proliferation of mouse HT - 2 cells is ≤ 0.05 ng/mL, corresponding to a specific activity of ≥ 2 x 107 units/mg.
  • Protein Synonyms:
    Tumor necrosis factor ligand superfamily member 10|Apo-2 ligand|Apo-2L|TRAIL|CD253|TL2|APO2L
  • Protein/Peptide Name:
    TGF beta 1
  • Purity:
    >98%
  • Formulation:
    Lyophilized

Specifications

Related Information