Order Entry
Puerto Rico
ContactUsLinkComponent
ELISA Assays for Infectious Disease and Autoimmune, Gold Standard Diagnostics
ELISA Assays for Infectious Disease and Autoimmune, Gold Standard Diagnostics
Certificates
undefined
ELISA Assays for Infectious Disease and Autoimmune, Gold Standard Diagnostics

Specifications

  • Target protein:
    Anti-Jo-1
  • Sample Type:
    Serum

Specifications

Related Information

Frequently Bought Together