Specifications
- Antibody Type:Primary
- Antigen Name:tryptophan hydroxylase 2
- Antigen Symbol:TPH2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- ImmunoChemistry:Yes
- Reactivity:Rat
- Western Blot:Yes
- Size:0.1 mL
- Form:Liquid
- Gene ID:P17290
- Antigen Synonyms:TPH2
- Storage Buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular Weight:55
- Storage Temperature:For long term storage –80 °C is recommended, but shorter term storage at –20 °C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
- Shipping Temperature:4 °C
- Immunogen:TPH polyclonal antibody was raised against recombinant rabbit tryptophan hydroxylase, isolated as inclusion bodies from E.
- Purification:Affinity Purified
- Cat. No.:10075-550
- Supplier no.:XPS-2030
Specifications
About this item
Tryptophan hydroxylase (TPH) catalyzes the first step in the biosynthesis of serotonin and melatonin. In mammals, serotonin biosynthesis occurs predominantly in neurons which originate in the raphe nuclei of the brain, and melatonin synthesis takes place within the pineal gland. Although TPH catalyzes the same reaction within the raphe nuclei and the pineal gland, TPH activity is rate-limiting for serotonin but not melatonin biosynthesis..
Tryptophan Hydroxylase antibody can be used for specific immunolabeling of the ~55k TPH protein in Western blots of human dorsal Raphe nucleus. Applications include Dot Blots (DB) Immunohistochemistry (IHC) and Western Blots (WB). The antibody has been used successfully with rat and human brain tissues in blot immunolabeling and immunohistochemical procedures. Based upon the relatively high degree of homology of tryptophan hydroxylase, the antibodies should cross-react with other mammalian species. When internally tested under ideal conditions the working dilutions were 1:1000 for IHC, DB and WB.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
For long term storage –80°C is recommended, but shorter term storage at –20°C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
Type: Primary
Antigen: TPH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Rat