- Antibody Type:Primary
- Antigen Name:synapsin I
- Antigen Symbol:SYN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:10 ug
- Form:Lyophilized
- Gene ID:P17599
- Antigen Synonyms:SYN1
- Molecular Weight:78
- Storage Temperature:For long term storage –80 °C is recommended, but shorter term storage at –20 °C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
- Shipping Temperature:RT
- Immunogen:Synapsin 1 polyclonal antibody was raised against synapsin I native protein purified from bovine brain.
- Purification:Affinity Purified
- Cat. No.:10075-702
- Supplier no.:XPS-2029
Synapsin I plays a key role in synaptic plasticity in brain. This effect is due in large part to the ability of the synapsins to regulate the availability of synaptic vesicles for release. In addition to its role in plasticity, the expression of synapsin I is a precise indicator of synapse formation. Thus synapsin I immunocytochemistry provides a valuable tool for the study of synaptogenesis. The role of synapsin in synaptic plasticity and in synaptogensis is regulated by phosphorylation.
Synapsin I antibody can be used for strong and specific immunolabeling of the Synapsin I 78k doublet in Western blots of rat brain. Immunolabeling blocked by preadsorption of antibody with Synapsin. Applications include Dot Blots (DB), Western Blots (WB), Immunohistochemistry (IHC), Immunofluorescence (IF) and Immunoprecipitation (IP). Human, mouse and rat have 100% amino acid sequence for Synapsin I. When internally tested under ideal conditions the working dilutions were 1:1000 for DB and WB, 1:2000 for IHC and IF, and for IP 1 ?g will quantitatively immunoprecipitate all synapsin in 200 ?g rat brain.
Affinity Purified
For long term storage –80°C is recommended, but shorter term storage at –20°C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
Type: Primary
Antigen: SYN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat