Specifications
- Antibody Type:Primary
- Antigen Name:gamma-aminobutyric acid (GABA) A receptor, gamma 2
- Antigen Symbol:Gabrg2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Bovine,Rat
- Western Blot:Yes
- Size:.05 mL
- Form:Liquid
- Gene ID:P18508
- Antigen Synonyms:Gabrg2
- Molecular Weight:46
- Storage Temperature:GABA A Receptor gamma 2 antibody can be stored at –20 °C and is stable at –20 °C for at least 1 year.
- Shipping Temperature:4 °C
- Immunogen:GABAA Receptor y2 polyclonal antibody was raised against a peptide representing a sequence that is specific for the gamma2 subunit of the receptor.
- Purification:Neat Serum
- Cat. No.:10075-356
- Supplier no.:XPS-2013
Specifications
About this item
Gamma-aminobutyric acid (GABA) is the primary inhibitory neurotransmitter in the central nervous system, causing a hyperpolarization of the membrane through the opening of a Cl- channel associated with the GABAA-Receptor (GABAA-R) subtype. GABAA-Rs are important therapeutic targets for a range of sedative, anxiolytic, and hypnotic agents and are implicated in several diseases including epilepsy, anxiety, depression, and substance abuse. The GABAA-R is a multimeric subunit complex. To date six alphas, four betas and four gammas, plus alternative splicing variants of some of these subunits, have been identified. Injection in oocytes or mammalian cell lines of cRNA coding for alpha and beta subunits results in the expression of functional GABAA-Rs sensitive to GABA. However, coexpression of a gamma subunit is required for benzodiazepine modulation. The various effects of the benzodiazepines in brain may also be mediated via different alpha subunits of the receptor. Lastly, phosphorylation of beta subunits of the receptor has been shown to modulate GABAA-R function.
GABAA Receptor antibody recognizes 45 - 47 k band in Western blots of rat brain membranes. Peptide antigen has no homology with any other GABAA-R or ? or ? subunits. Applications include Dot Blots (DB) and Western Blots (WB). Bovine, human, mouse, and rat have 100% amino acid sequence identity with the antigen used to raise the antibody. When internally tested under ideal conditions the working dilutions were 1:1000 for DB and WB.
Neat Serum
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: Gabrg2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat, Bovine