- Antibody Type:Primary
- Antigen Name:tyrosine hydroxylase
- Antigen Symbol:Th
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:0.1 mL
- Form:Liquid
- Gene ID:P04177
- Antigen Synonyms:Th
- Modification:Ser 31
- Storage Buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular Weight:60
- Storage Temperature:For long term storage –80 °C is recommended, but shorter term storage at –20 °C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
- Shipping Temperature:4 °C
- Immunogen:TH (Ser31) polyclonal antibody was raised against a synthetic phosphopeptide corresponding to amino acids residues surrounding the phospho-Ser31 of rat tyrosine hydroxylase (TH).
- Purification:Affinity Purified
- Cat. No.:10075-630
- Supplier no.:XPS-1032
Tyrosine Hydroxylase (TH) is the rate-limiting enzyme in the synthesis of the catecholamines Dopamine and Norepinephrine. TH antibodies can therefore be used as markers for dopaminergic and noradrenergic neurons. We raised this polyclonal antibody against a peptide representing the sequence around Ser19 in rat TH purified. This antibody is suitable for most immunochemical applications in a variety of mammalian and some non-mammalian species.
Applications include Dot Blots (DB), Immunoblotting (IB), Immunofluorescence (IF), Immunohistochemistry (IHC) and Western Blots (WB). Mouse and rat have 100% amino acid sequence identity with the antigen used to raise the antibody. When internally tested under ideal conditions the working dilutions were 1:1000 for IF, IHC, DB and WB.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
For long term storage –80°C is recommended, but shorter term storage at –20°C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
Type: Primary
Antigen: Th
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat