Specifications
- Antibody Type:Primary
- Antigen Name:mitogen-activated protein kinase kinase 1
- Antigen Symbol:MAP2K1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Xenopus,Human,Rat,Mouse
- Western Blot:Yes
- Size:0.1 mL
- Form:Liquid
- Gene ID:Q02750
- Antigen Synonyms:MAP2K1
- Modification:Ser 218/Ser 222
- Storage Buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular Weight:45
- Storage Temperature:For long term storage –80 °C is recommended, but shorter term storage at –20 °C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
- Shipping Temperature:4 °C
- Immunogen:MEK 1/2 (Ser218/222) polyclonal antibody was raised against a synthetic phosphopeptide corresponding to amino acids residues surrounding the phosphoSer218/222 of human MEK 1/2.
- Purification:Affinity Purified
- Cat. No.:10075-616
- Supplier no.:XPS-1018
Specifications
About this item
MEK 1 and MEK 2 are integral components of the MAP kinase cascade that regulates cell growth and differentiation and this pathway also plays a key role in synaptic plasticity in brain. Activation of MEK 1/2 occurs via phosphorylation of two serine residues (Ser218 and Ser222). Activated MEK 1/2 then acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on ERK. This phosphorylation of ERK by MEK 1/2 is a critical step in the MAP kinase cascade.
The antibody immunolabels the ~45k MEK protein in a Western blot of serum-stimulated NIH 3T3 cells. The immunolabeling is blocked by the phosphopeptide used as antigen but not the corresponding non-phosphopeptide. Applications include Dot Blots (DB) and Western Blots (WB). Suitability for Immunohistochemistry (IHC) has not yet been determined. Human, mouse, rat and Xenopus have 100% amino acid sequence identity with the antigen used to raise the antibody. When internally tested under ideal conditions the working dilutions were 1:1000 for DB and WB.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
For long term storage –80°C is recommended, but shorter term storage at –20°C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
Type: Primary
Antigen: MAP2K1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Xenopus