Specifications
- Antibody Type:Primary
- Antigen Name:mitogen-activated protein kinase kinase 1
- Antigen Symbol:MAP2K1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Rat
- Western Blot:Yes
- Size:0.1 mL
- Form:Liquid
- Gene ID:Q02750
- Antigen Synonyms:MAP2K1
- Modification:Thr 386
- Storage Buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular Weight:45
- Storage Temperature:For long term storage –80 °C is recommended, but shorter term storage at –20 °C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
- Shipping Temperature:4 °C
- Immunogen:MEK 1 (Thr386) polyclonal antibody was raised against a synthetic phosphopeptide corresponding to amino acids residues surrounding the phospho Thr386 of human MEK 1.
- Purification:Affinity Purified
- Cat. No.:10075-536
- Supplier no.:XPS-1017
Specifications
About this item
MEK 1 (MAP Kinase Kinase, also known as MKK) is an integral component of the MAP kinase cascade that regulates cell growth and differentiation and this pathway also plays a key role in synaptic plasticity in brain. Activated MEK 1 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase. Conversely there also appears to be a feedback phosphorylation of MEK 1 by MAP kinase. The sites on MEK 1 that are phosphorylated by MAP kinase are Thr292 and Thr386.
Applications include Dot Blots (DB) and Western Blots (WB). Suitability for Immunohistochemistry (IHC) has not yet been determined. When internally tested under ideal conditions the working dilutions were 1:1000 for DB and WB.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
For long term storage –80°C is recommended, but shorter term storage at –20°C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
Type: Primary
Antigen: MAP2K1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat