Specifications
- Antibody Type:Primary
- Antigen Name:myristoylated alanine rich protein kinase C substrate
- Antigen Symbol:Marcks
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Xenopus,Human,Rat,Mouse
- Western Blot:Yes
- Size:0.1 mL
- Form:Liquid
- Gene ID:P30009
- Antigen Synonyms:Marcks
- Modification:Ser 152/156
- Storage Buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular Weight:87
- Storage Temperature:For long term storage –80 °C is recommended, but shorter term storage at –20 °C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
- Shipping Temperature:4 °C
- Immunogen:MARCKS (Ser152/156) polyclonal antibody was raised against a synthetic phosphopeptide corresponding to amino acid residues surrounding the phospho-Ser152 and phospho-Ser156 of rat MARCKS.
- Purification:Affinity Purified
- Cat. No.:10075-666
- Supplier no.:XPS-1015
Specifications
About this item
Myristoylated Alanine-Rich C Kinase Substrate (MARCKS) is a major substrate for phosphorylation by protein kinase C (PKC). The phosphorylation of Ser152/156 can be used as a measure of PKC activation. Moreover, MARCKS is a member of a family of calmodulin binding proteins. Phosphorylation of Ser152/156 modulates the binding of MARCKS to calmodulin.
Immunolabeling of the 87kDa band in rat brain homogenate is blocked by the Ser152/156 phosphopeptide but not by the non-phosphopeptide. The antibody is purified by sequential chromatography on phospho- and non-phosphopeptide affinity columns. Antibody dilutions and tissue load should be based on tissue type and expected phosphorylation state. Initial recommended range of dilutions: 1:500 to 1:2000. Applications include Dot Blots (DB) and Western Blot (WB). Suitability for Immunohistochemistry (IHC) not yet determined. Human, mouse, rat, and Xenopus have 100% amino acid sequence identity with the antigen used to raise the antibody. When internally tested under ideal conditions the working dilutions were 1:1000 for DB and WB.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
For long term storage –80°C is recommended, but shorter term storage at –20°C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
Type: Primary
Antigen: Marcks
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Xenopus