Specifications
- Antibody Type:Primary
- Antigen Name:interferon (alpha, beta and omega) receptor 1
- Antigen Symbol:IFNAR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Bovine,Rat,Mouse
- Western Blot:Yes
- Size:0.1 mL
- Form:Liquid
- Gene ID:P17181
- Antigen Synonyms:IFNAR1
- Modification:Ser 535/539
- Storage Buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular Weight:110-130
- Storage Temperature:For long term storage –80 °C is recommended, but shorter term storage at –20 °C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
- Shipping Temperature:4 °C
- Immunogen:IFNAR1 (Ser535/539) polyclonal antibody was raised against a synthetic phosphopeptide corresponding to amino acid residues surrounding the human Phospho-Ser535, 539 Type I Interferon-a Receptor Subunit 1 (IFNAR1).
- Purification:Affinity Purified
- Cat. No.:10075-388
- Supplier no.:XPS-1014
Specifications
About this item
Interferons are widely used therapeutic agents because of their anti tumor and antiviral effects and because of their modulatory effects on the immune system. These cytokines produce their effects by binding to the Type 1 Interferon-alpha Receptor (IFNAR1). Down regulation of this receptor plays a key role in determining the magnitude and duration of cytokine signaling. This down regulation is thought to be influenced by phosphorylation of Serine 535 and 539 in the IFNAR1.
Applications include Dot Blots (DB) and Western Blots (WB). Suitability for Immunohistochemistry (IHC) has not yet been determined. When internally tested under ideal conditions the working dilutions were 1:1000 for DB and WB.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
For long term storage –80°C is recommended, but shorter term storage at –20°C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
Type: Primary
Antigen: IFNAR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Bovine