Specifications
- Antibody Type:Primary
- Antigen Name:calcium/calmodulin-dependent protein kinase II inhibitor 2
- Antigen Symbol:CAMK2N2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:P11798
- Antigen Synonyms:CAMK2N2
- Storage Buffer:Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Molecular Weight:50 kDa
- Storage Temperature:Store antibody at –20 °C for up to one year.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:CaMKII (Ab-286) antibody was raised against a peptide sequence around aa.284~288 (Q-E-T-V-D) derived from Human CaMKII.
- Purification:Antibodies were purified by affinity-chromatography using epitope-specific peptide.
- Cat. No.:10069-018
- Supplier no.:79-605
Specifications
About this item
CaM-kinase II (CAMK2) is a prominent kinase in the central nervous system that may function in long-term potentiation and neurotransmitter release. Member of the NMDAR signaling complex in excitatory synapses it may regulate NMDAR-dependent potentiation of the AMPAR and synaptic plasticity
Western Blot: 1:500~1:1000, Immunofluorescence: 1:100~1:200
Antibodies were purified by affinity-chromatography using epitope-specific peptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: CAMK2N2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat