Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-STAT5A Rabbit Polyclonal Antibody
Anti-STAT5A Rabbit Polyclonal Antibody
Catalog # 10068-804
Supplier:  Prosci
undefined
Anti-STAT5A Rabbit Polyclonal Antibody
Catalog # 10068-804
Supplier:  Prosci
Supplier Number:  79-289

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    signal transducer and activator of transcription 5A
  • Antigen Symbol:
    STAT5A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    0.1 mg
  • Form:
    Liquid
  • Gene ID:
    P42229
  • Antigen Synonyms:
    STAT5A
  • Storage Buffer:
    Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
  • Molecular Weight:
    90 kDa
  • Storage Temperature:
    Store antibody at –20 °C for up to one year.
  • Concentration:
    1 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    STAT5A (Ab-694) antibody was raised against a peptide sequence around aa. 692~696 (D-G-Y-V-K) derived from Human STAT5A.
  • Purification:
    Antibodies were purified by affinity-chromatography using epitope-specific peptide.
  • Cat. No.:
    10068-804
  • Supplier no.:
    79-289

Specifications

About this item

Carries out a dual function: signal transduction and activation of transcription. Binds to the GAS element and activates PRL-induced transcription.

Western Blot: 1:500~1:1000, Immunohistochemistry: 1:50~1:100

Antibodies were purified by affinity-chromatography using epitope-specific peptide.

Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: STAT5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat