- Antibody Type:Primary
- Antigen Name:RVFV_sL_gp1 polymerase [ Rift Valley fever virus ]
- Antigen Symbol:RVFV_sL_gp1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Virus
- Size:0.1 mg
- Form:Liquid
- Gene ID:A2SZS2
- Antigen Synonyms:RVFV_sL_gp1
- Storage Buffer:RVF virus Polymerase Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular Weight:Predicted: 230 kDa
- Storage Temperature:Rift Valley Fever Virus Polymerase antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:RVF virus Polymerase antibody was raised against a 16 amino acid peptide near the amino terminus of the RVF virus Polymerase. The immunogen is located within amino acids 370 - 420 of Rift Valley Fever Virus Polymerase.
- Purification:Rift Valley Fever Virus Polymerase Antibody is affinity chromatography purified via peptide column.
- Cat. No.:10061-922
- Supplier no.:7415
Rift Valley Fever Virus Polymerase Antibody: Rift Valley Fever (RFV) virus is an arthropod-borne virus endemic to Africa that infects humans and animals that is transmitted predominantly by mosquitoes. During human infections, symptoms can range from benign fever to severe encephalitis and fatal hepatitis with hemorrhagic fever. The Bunyaviridae family of viruses to which the RVF virus belongs are spherical enveloped viruses with a tripartite RNA genome of negative or ambisense polarity. The three segments are referred to as the L, M, and S segments. The L and M segments are negative polarity and code fore the L-dependent RNA polymerase and glycoprotein precursor respectively. The S segment is of ambisense polarity and encodes the nucleoprotein and non-structural proteins.
RVF virus Polymerase antibody can detect 10 ng RVF virus Polymerase peptide in ELISA at 1 ?g/mL.
Rift Valley Fever Virus Polymerase Antibody is affinity chromatography purified via peptide column.
RVF virus Polymerase Antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: RVFV_sL_gp1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus